missing translation for 'onlineSavingsMsg'
Learn More

Zinc peroxide, approximately 50% ZnO{2}, Thermo Scientific Chemicals

Curing agent, rubber accelerator

£36.88 - £110.55

Chemical Identifiers

CAS 1314-22-3
Molecular Formula O2Zn
Molecular Weight (g/mol) 97.38
MDL Number MFCD00137469
InChI Key IPTOGCUGCFHDSS-UHFFFAOYSA-N
Synonym zinc peroxide, zinc superoxide, zinc dioxide, zinc peroxide un1516 oxidizer, zinc peroxide, medicinal, unii-0i969dvm77, zinc peroxide, medicinal usp, zpo
PubChem CID 14809
SMILES [Zn++].[O-][O-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15431547
View Documents
Thermo Scientific Alfa Aesar
039635.22
100 g
£36.88
100g
Please to purchase this item. Need a web account? Register with us today!
15441547
View Documents
Thermo Scientific Alfa Aesar
039635.36
500 g
£110.55
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zinc peroxide is used as curing agent, rubber accelerator. has been applied in a variety of settings, from medicine to aesthetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-22-3
97.38
IPTOGCUGCFHDSS-UHFFFAOYSA-N
14809
O2Zn
MFCD00137469
zinc peroxide, zinc superoxide, zinc dioxide, zinc peroxide un1516 oxidizer, zinc peroxide, medicinal, unii-0i969dvm77, zinc peroxide, medicinal usp, zpo
[Zn++].[O-][O-]

Specifications

>150°C (decomposition)
Powder
MFCD00137469
14,10149
Insoluble in water.
[Zn++].[O-][O-]
97.38
97.38
Odorless
1314-22-3
O2Zn
UN1516
zinc peroxide, zinc superoxide, zinc dioxide, zinc peroxide un1516 oxidizer, zinc peroxide, medicinal, unii-0i969dvm77, zinc peroxide, medicinal usp, zpo
IPTOGCUGCFHDSS-UHFFFAOYSA-N
zinc(2+) dioxidanediide
14809
≈50% ZnO2
Zinc peroxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H332-H315-H319-H335
May intensify fire
oxidizer.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P280-P283-P306+P360-P370+P378q-P371+P380+P375-P501c

H271

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: ZINC PEROXIDE

missing translation for 'einecsNumber' : 215-226-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only