missing translation for 'onlineSavingsMsg'
Learn More

Zinc phosphide, 99.999% (metals basis)

Product Code. p-7059844
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
This item is not returnable. View return policy

Product Code. 15452277

Brand: Thermo Scientific Alfa Aesar 045459.02

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-84-7 | Zn3P2

Zinc phosphide has useful applications in photovoltaic cells. It is also employed as a laboratory chemical. It plays an important role in medicine and pharmaceuticals. It acts as a fumigant in grain storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-84-7
Molecular Formula Zn3P2
MDL Number MFCD00049631

Specifications

Boiling Point 1100°C
Melting Point 420°C
Physical Form Pieces
Quantity 0.5 g
UN Number UN1714
Merck Index 14,10152
Solubility Information Insoluble in water and ethanol. Soluble in benzene.
Formula Weight 258.1
Percent Purity 99.999%
Sensitivity Moisture sensitive
Density 4.55 g/mL
Assay (metals basis)
Chemical Name or Material Zinc phosphide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.