Learn More
R&L Enterprises™ Zinc Plated Stand Rods
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Zinc Plated Rods are manufactured from mild steel.
Brand:  R&L Enterprises™ 3108/11
 View more versions of this product
Product Code. 12358139
  • £115.83 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Threaded
  • Available for a range of diameters and lengths
  • Length of rod does not include the thread
Specifications
Specifications
Zinc Plated Mild Steel
500 mm
10 mm
Rod
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
R&L Enterprises™ Zinc Plated Stand Rods > Length: 500mm; Diameter: 10mm