missing translation for 'onlineSavingsMsg'
Learn More

Zinc powder, -100 mesh, 97+%, Thermo Scientific Chemicals

Product Code. 11650222
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11650222

Brand: Thermo Scientific Alfa Aesar L13310.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zinc powder is used in thermal spraying for corrosion protection and in the chemical industry. It provides chemical potential energy in alkaline batteries to pump electrical charges between the negative and positive terminals. It is used as a fuel for model rockets with a mixture of 2.04 parts zinc to one part sulfur. It is used as protective pigment in industrial coatings, as a reducing agent and catalyst in a variety of chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc powder is used in thermal spraying for corrosion protection and in the chemical industry. It provides chemical potential energy in alkaline batteries to pump electrical charges between the negative and positive terminals. It is used as a fuel for model rockets with a mixture of 2.04 parts zinc to one part sulfur. It is used as protective pigment in industrial coatings, as a reducing agent and catalyst in a variety of chemical reactions.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to air and moisture. Incompatible with strong acids, strong bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Spezifikation

Melting Point 420°C
Boiling Point 907°C
Density 7.13
Quantity 100 g
Mesh Size -100 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
UN Number UN1436
Solubility Information Reacts with water.
Percent Purity ≥97%
Chemical Name or Material Zinc powder
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Zinc powder
Signalwort
  • Danger
Gefahrenkategorie
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Gefahrenhinweis
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231-Handle under inert gas.
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Ergänzende Informationen
  • MIXTURE LIST-Contains : zinc powder

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt