missing translation for 'onlineSavingsMsg'
Learn More

Zinc rod, 7mm (0.3 in.) dia., 99.9985% (metals basis), Thermo Scientific Chemicals

Product Code. 11307137
Click to view available options
Quantity:
100 mm
5 x 100 mm

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Quantity 100 mm
Percent Purity 99.9985%
Odor Odorless
Weight ≈2.74g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Zinc rod, 7mm (0.3 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'