missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™
Click to view available options
Quantity:
30 mL
250 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Zn | 1000μg/mL |
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Packaging | Triple-leached LDPE bottle |
CAS Min % | 1.0, 5.0, 94.0 |
Quantity | 30 mL |
CAS Max % | 1.0, 5.0, 94.0 |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Grade | Plasma |
Concentration or Composition (by Analyte or Components) | Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLZN2-2Y, -2T, -2X 10,000 PLZN2-3Y, -3X 2-5% |
Solvent or Matrix | 2% HNO3 |
Chemical Name or Material | Zinc Standard |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- 1000µg/ml zinc
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- MIXTURE LIST-Contains : nitric acid, zinc
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'