missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
£59.25
Chemical Identifiers
CAS | 7779-88-6 |
---|---|
Molecular Formula | N2O6Zn |
Molecular Weight (g/mol) | 189.388 |
MDL Number | 11291 |
InChI Key | ONDPHDOFVYQSGI-UHFFFAOYSA-N |
Synonym | zinc nitrate, celloxan, zinc dinitrate, nitric acid, zinc salt, unii-edo66f5u49, nitric acid, zinc salt 2:1, nitrate de zinc, x 4 nitrate, nitrate de zinc french, zinc nitrate zn no3 2 |
PubChem CID | 24518 |
IUPAC Name | zinc;dinitrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Zn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10408990
|
Fisher Chemical
J/8070/08 |
500 mL | N/A | ||||||
Description
Chemical Identifiers
7779-88-6 | |
189.388 | |
ONDPHDOFVYQSGI-UHFFFAOYSA-N | |
24518 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Zn+2] |
N2O6Zn | |
11291 | |
zinc nitrate, celloxan, zinc dinitrate, nitric acid, zinc salt, unii-edo66f5u49, nitric acid, zinc salt 2:1, nitrate de zinc, x 4 nitrate, nitrate de zinc french, zinc nitrate zn no3 2 | |
zinc;dinitrate |
Specifications
Colorless | |
7779-88-6 | |
11291 | |
ONDPHDOFVYQSGI-UHFFFAOYSA-N | |
zinc;dinitrate | |
24518 | |
Liquid |
1 | |
N2O6Zn | |
zinc nitrate, celloxan, zinc dinitrate, nitric acid, zinc salt, unii-edo66f5u49, nitric acid, zinc salt 2:1, nitrate de zinc, x 4 nitrate, nitrate de zinc french, zinc nitrate zn no3 2 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Zn+2] | |
189.388 | |
HDPE plastic bottle |