missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 557-05-1 |
---|---|
MDL Number | MFCD00013031 |
InChI Key | XOOUIPVCVHRTMJ-UHFFFAOYSA-L |
Synonym | zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat |
PubChem CID | 11178 |
IUPAC Name | zinc;octadecanoate |
SMILES | CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10018511
|
Thermo Scientific Acros
263780010 |
1 kg |
£27.54
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10202512
|
Thermo Scientific Acros
263780050 |
5 kg |
£89.14
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10731804
|
Thermo Scientific Acros
263780200 |
20 kg |
£273.36
20kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
557-05-1 | |
XOOUIPVCVHRTMJ-UHFFFAOYSA-L | |
11178 | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2] |
MFCD00013031 | |
zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat | |
zinc;octadecanoate |
Specifications
557-05-1 | |
White | |
Authentic | |
C36H70O4Zn | |
MFCD00013031 | |
zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat | |
XOOUIPVCVHRTMJ-UHFFFAOYSA-L | |
zinc;octadecanoate | |
11178 | |
12.5 to 14.0% (ZnO) | |
Zinc stearate |
118.0°C to 128.0°C | |
>100°C | |
Plastic bottle | |
[CH3(CH2)16CO2]2Zn | |
15, 10358 | |
Solubility in water: insoluble. Other solubilities: insoluble in ethanol,diethyl ether,,soluble in benzene,xylene,fats,oils | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2] | |
632.34 | |
632.34 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 209-151-9
missing translation for 'rtecsNumber' : ZH5200000
missing translation for 'tsca' : TSCA
RUO – Research Use Only