Learn More
Zinc sulfate heptahydrate, 99%, ACS reagent, Thermo Scientific Chemicals
£34.10 - £245.00
Chemical Identifiers
CAS | 7446-20-0 |
---|---|
Molecular Formula | H14O11SZn |
Molecular Weight (g/mol) | 287.54 |
MDL Number | MFCD00149894 |
InChI Key | RZLVQBNCHSJZPX-UHFFFAOYSA-L |
Synonym | zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o |
PubChem CID | 62640 |
ChEBI | CHEBI:32312 |
SMILES | O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10065850
|
Thermo Scientific Acros
424605000 |
500 g | Plastic bottle |
£34.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10267273
|
Thermo Scientific Acros
424600010 |
1 kg | Plastic bottle |
£54.80
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10301461
|
Thermo Scientific Acros
424600030 |
3 kg | Plastic bottle |
£132.00
3kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10013633
|
Thermo Scientific Acros
424600100 |
10 kg | Plastic drum |
£245.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7446-20-0 | |
287.54 | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
62640 | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
H14O11SZn | |
MFCD00149894 | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
CHEBI:32312 |
Specifications
100.0°C | |
White | |
Crystals | |
H14O11SZn | |
MFCD00149894 | |
zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
zinc;sulfate;heptahydrate | |
62640 | |
287.53 | |
ACS Reagent |
7446-20-0 | |
4.4 to 6.0 (5% soln. at 25°C) | |
99.0 to 103% (Complexometry) | |
ZnSO4·7H2O | |
15, 10359 | |
Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol, insoluble in alcohol | |
O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
287.54 | |
CHEBI:32312 | |
99% | |
Zinc sulfate heptahydrate |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : ZH5300000
- Zinc sulfate heptahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Zinc sulfate heptahydrate, Zinc sulfate
RUO – Research Use Only