missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate Heptahydrate, Certified AR for Analysis, 99.5-100.5%, Fisher Chemical™
55.22 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
H14O11SZn, CAS Number-7446-20-0, zinc sulfate heptahydrate, znso4.7h2o, zinc sulfate jan, zinc sulfate znso4 heptahydrate, white vitriol heptahydrate, ccris 5563, zinc sulfate heptahydrate 1:1:7, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, 500g, CHEBI:32312, White
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 100°C |
Color | White |
pH | 4.4 to 6 |
Physical Form | Solid |
Quantity | 500 g |
IUPAC Name | zinc(2+) heptahydrate sulfate |
Formula Weight | 287.53g/mol |
Packaging | Plastic powder jar |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Zinc sulfate heptahydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'