missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate Monohydrate, ≥99.9% trace metals basis, Honeywell™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 238°C |
Quantity | 100 g |
Linear Formula | ZnSO4 · H2O |
UN Number | UN3077 |
Solubility Information | Soluble in water |
Formula Weight | 179.47g/mol |
Percent Purity | ≥99.9% (trace metals basis) |
Packaging | Poly Bottle |
Chemical Name or Material | Zinc Sulfate Monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'