missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate monohydrate, 99%, Thermo Scientific Chemicals

Product Code. 10559993
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-19-7
Molecular Formula O4SZn·H2O
Molecular Weight (g/mol) 179.47
MDL Number MFCD00149893
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
PubChem CID 62639
IUPAC Name zinc;sulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[Zn+2]

Specifications

Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula ZnSO4·H2O
Merck Index 15, 10359
Solubility Information Solubility in water: 350g/L (20°C). Other solubilities: practically insoluble in ethanol
Formula Weight 179.47
Percent Purity 99%
Identification Confirmed (Zinc), Confirmed (Sulfate)
Packaging Plastic bottle
Chemical Name or Material Zinc sulfate monohydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc sulfate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.