missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11379239
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Zinc sulfide is used in luminescent materials, infra red optical materials, semiconductors, as a pigment, and as a window for visible optics and infrared optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-98-3
Molecular Formula SZn
Molecular Weight (g/mol) 97.44
MDL Number MFCD00011301
InChI Key DRDVZXDWVBGGMH-UHFFFAOYSA-N
Synonym zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
PubChem CID 14821
SMILES [S--].[Zn++]

Specifications

Melting Point 1700°C
Physical Form Pieces
Quantity 10 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in acids. Insoluble in water.
IUPAC Name sulfanylidenezinc
Formula Weight 97.43
Density 4.1
Chemical Name or Material Zinc Sulfide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'