missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulphate 0.1 M According to European Pharmacopoeia (EP), Reagecon
BIOPHARMA

Fully factorized, high-purity, stable product developed and tested for titrations.

Brand:  Reagecon 3008600


Product Code. 11478879

  • £83.35 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
  • Accurate, stable, reproducible and fully factorized.
  • Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
  • Stability is proven (either opened or unopened) for the shelf life of this product.
  • Certified to a specification of ± 0.2%.
  • Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
  • May be used for manual titrations, or automatic titrations, to determine the end points by detection as detailed in several EP monographs.
  • NIST traceable.
Specifications

Specifications

Accuracy ± 0.2%
HDPE Bottle GL45
Titration and EP Monographs
0.1 M
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : zinc sulphate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers