missing translation for 'onlineSavingsMsg'
Learn More
Zinc telluride, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11448610

Zinc telluride, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11448610
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11448610

Brand: Thermo Scientific Alfa Aesar 044412.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zinc telluride can be easily doped, and for this reason it is one of the more common semiconducting materials used in optoelectronics. ZnTe is important for development of various semiconductor devices, including blue LEDs, laser diodes, solar cells, and components of microwave generators. It can be used for solar cells, for example, as a back-surface field layer and p-type semiconductor material for a CdTe/ZnTe structure[6] or in PIN diode structures. The material can also be used as a component of ternary semiconductor compounds.Zinc telluride together with lithium niobate is often used for generation of pulsed terahertz radiation in time-domain terahertz spectroscopy and terahertz imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-11-3
Molecular Formula TeZn
Molecular Weight (g/mol) 192.98
MDL Number MFCD00043079
InChI Key NSRBDSZKIKAZHT-UHFFFAOYSA-N
Synonym zinc telluride, zinc telluride znte, unii-ir8eb6g3vq, zinktellurid, ir8eb6g3vq, zinc telluride,-100 mesh trace metals basis
PubChem CID 3362486
IUPAC Name tellanylidenezinc
SMILES [Zn]=[Te]

Specifications

Melting Point 1240°C
Color Red
Physical Form Powder
Quantity 5 g
Merck Index 14,10162
Solubility Information It decomposes in water.
Formula Weight 192.97
Percent Purity 99.99%
Odor Odorless
Refractive Index 3.56
Density 5.9 g/mL
Assay (metals basis)
Chemical Name or Material Zinc telluride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ17K010 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog Number044412.22,044412.06,044412.14
Lot Number10253194 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number044412.06,044412.14
Lot NumberZ05I042 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog Number044412.06,044412.14
Lot NumberX04I048 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number044412.06,044412.14
Lot NumberU27H043 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044412.06,044412.14
compliance-icons
Product Identifier
  • Zinc telluride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Zinc telluride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc telluride, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.