missing translation for 'onlineSavingsMsg'
Learn More

Zinc telluride, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11358038
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Zinc telluride is a semiconductor material used in optoelectronics. It is used in blue LEDs, laser diodes, solar cells, components of microwave generators, electro-optic detector, holographic interferometry and laser optical phase conjugation devices. It is associated with lithium niobate and used for the generation of pulsed terahertz radiation in time-domain terahertz spectroscopy and terahertz imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-11-3
Molecular Formula TeZn
Molecular Weight (g/mol) 192.98
MDL Number MFCD00043079
InChI Key NSRBDSZKIKAZHT-UHFFFAOYSA-N
Synonym zinc telluride, zinc telluride znte, unii-ir8eb6g3vq, zinktellurid, ir8eb6g3vq, zinc telluride,-100 mesh trace metals basis
PubChem CID 3362486
IUPAC Name tellanylidenezinc
SMILES [Zn]=[Te]

Specifications

Melting Point 1240°C
Color Red
Physical Form Lump
Quantity 50 g
Assay Percent Range 99.998% metal basis
Solubility Information Insoluble in water.
Formula Weight 192.97
Odor Odorless
Refractive Index 3.56
Density 6.34
Chemical Name or Material Zinc telluride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.