missing translation for 'onlineSavingsMsg'
Learn More
Zinc tert-butoxide, Thermo Scientific Chemicals Product Code.: 11328648

Zinc tert-butoxide, Thermo Scientific Chemicals

Product Code. 11328648
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11328648

Brand: Thermo Scientific Alfa Aesar 040718.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A sol-gel process has been developed for the chemical synthesis of ZnS at room temperature using zinc tert-butoxide and H 2 S as precursors in a toluene solution. The key step that enables this zinc-mediated epoxidation to be catalytic may be the equilibrium between zinc peroxide (R)-15 and zinc tert-butoxide (R)-16 in the presence of t-BuOOH. An additional important feature that renders the dual catalytic cycles feasible is the poor reactivity of zinc tert-butoxide toward the iridium-bound cationic intermediate, whereby the requisite deprotonation process is effectively mediated by zinc tert-butoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4278-43-7
Molecular Formula C8H18O2Zn
Molecular Weight (g/mol) 211.61
MDL Number MFCD00145545
InChI Key DVGVEVPHJQJMPP-UHFFFAOYSA-N
Synonym zinc tert-butoxide, di-tert-butoxyzinc, acmc-20aj87, zinc bis 2-methylpropan-2-olate, zinc 2+ bis 2-methylpropan-2-olate, zinc 2+ ion bis 2-methylpropan-2-olateShow MoreShow Less
PubChem CID 14178434
IUPAC Name zinc;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].[Zn+2]

Specifications

Color White to Yellow
Odor Odorless
Linear Formula Zn[OC(CH3)3]2
Quantity 250 mg
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 211.61
Physical Form Powder
Chemical Name or Material Zinc tert-butoxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN27F029 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog Number040718.MD,040718.03
Lot NumberF02P15 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040718.MD,040718.03
Lot NumberB16S015 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040718.MD,040718.03
Lot NumberH08X037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040718.MD,040718.03
Lot NumberC21R048 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040718.MD,040718.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc tert-butoxide, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.