missing translation for 'onlineSavingsMsg'
Learn More

Zinc trifluoroacetate hydrate, Thermo Scientific Chemicals

£75.98 - £268.00

Chemical Identifiers

CAS 207801-31-8
Molecular Formula C4F6O4Zn
Molecular Weight (g/mol) 291.41
MDL Number MFCD00061595
InChI Key VCQWRGCXUWPSGY-UHFFFAOYSA-L
Synonym zinc trifluoroacetate hydrate, acmc-1ch94, zinc trifluoroacetate-water 1/2/1, zinc 2+ hydrate ditrifluoroacetate
PubChem CID 57375648
IUPAC Name zinc;2,2,2-trifluoroacetate;hydrate
SMILES [Zn++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363467
View Documents
Thermo Scientific Alfa Aesar
018686.14
25 g
£75.98
25g
Please to purchase this item. Need a web account? Register with us today!
11353467
View Documents
Thermo Scientific Alfa Aesar
018686.22
100 g
£268.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zinc trifluoroacetate hydrate is useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207801-31-8
291.41
VCQWRGCXUWPSGY-UHFFFAOYSA-L
57375648
[Zn++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
C4F6O4Zn
MFCD00061595
zinc trifluoroacetate hydrate, acmc-1ch94, zinc trifluoroacetate-water 1/2/1, zinc 2+ hydrate ditrifluoroacetate
zinc;2,2,2-trifluoroacetate;hydrate

Specifications

207801-31-8
MFCD00061595
Hygroscopic
Soluble in water.
[Zn++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
291.41
291.40 (Anhydrous)
Zinc trifluoroacetate hydrate
C4F6O4Zn
UN3077
zinc trifluoroacetate hydrate, acmc-1ch94, zinc trifluoroacetate-water 1/2/1, zinc 2+ hydrate ditrifluoroacetate
VCQWRGCXUWPSGY-UHFFFAOYSA-L
zinc;2,2,2-trifluoroacetate;hydrate
57375648
Chunks and/or Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only