missing translation for 'onlineSavingsMsg'
Learn More

Zinc wire, 0.25mm (0.01in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Form: approx. 5.60g/m

£109.55 - £293.46

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355457
View Documents
Thermo Scientific Alfa Aesar
042703.G5
5 m
£109.55
5m
Please to purchase this item. Need a web account? Register with us today!
11345457
View Documents
Thermo Scientific Alfa Aesar
042703.G9
25 m
£293.46
25m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
23994
zinc
Zn
MFCD00011291
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
CHEBI:30185
[Zn]

Specifications

Gray
Gray wire
Zn
dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
[Zn]
65.38
CHEBI:30185
Wire
7440-66-6
99.95% (Metals basis)
MFCD00011291
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
23994
Odorless
Zinc
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-175-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only