missing translation for 'onlineSavingsMsg'
Learn More

Zinc wire, 0.5mm (0.02in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11375457
Click to view available options
Quantity:
25 m
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
CAS 7440-66-6
Appearance Gray wire
Density 7.14g/cm3
Assay Percent Range 99.95% (Metals basis)
Molecular Formula Zn
MDL Number MFCD00011291
Quantity 25 m
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES [Zn]
IUPAC Name zinc
Molecular Weight (g/mol) 65.38
PubChem CID 23994
ChEBI CHEBI:30185
Odor Odorless
Form Wire
Chemical Name or Material Zinc
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'