Learn More
Merck ZipTip™ Pipet Tips
Sample preparation and precise liquid handling in one device
£57.70 - £4265.00
Specifications
Volume (Metric) | 10 μL |
---|---|
For Use With (Equipment) | Universal Fit |
Product Line | ZipTip |
Material | Polypropylene |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10300231
|
Merck
ZTC18S096 |
With C18 Media | 96/Pk. |
£631.00
Pack of 96 |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||||
10349141
|
Merck
ZTC04S008 |
With C4 Media | 8/Pk. |
£66.05
Pack of 8 |
Estimated Shipment: 15-10-2024
Log in to see stock. |
||||
10481253
|
Merck
ZTC04S096 |
With C4 Media | 96/Pk. |
£450.00
Pack of 96 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
10748354
|
Merck
ZTSCXS096 |
With Strong Cation Exchange Media | 96/Pk. |
£900.00
Pack of 96 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
10453382
|
Merck
ZTSCXS008 |
With Strong Cation Exchange Media | 8/Pk. |
£108.00
Pack of 8 |
Estimated Shipment: 12-12-2024
Log in to see stock. |
||||
10044590
|
Merck
ZTC18S960 |
With C18 Media | 960/Pk. |
£3665.00
Pack of 960 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
10093501
|
Merck
ZTC04S960 |
With C4 Media | 960/Pk. |
£4120.00
Pack of 960 |
Estimated Shipment: 11-11-2024
Log in to see stock. |
||||
10278851
|
Merck
ZTC18M960 |
With Micro Bed of C18 Media | 960/Pk. |
£4265.00
Pack of 960 |
Estimated Shipment: 12-12-2024
Log in to see stock. |
||||
10740883
|
Merck
ZTC18M096 |
With Micro Bed of C18 | 96/Pk. |
£514.00
Pack of 96 |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||||
10650852
|
Merck
ZTC18M008 |
With Micro Bed of C18 Media | 8/Pk. |
£66.50
Pack of 8 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
||||
10156390
|
Merck
ZTC18S008 |
With C18 Media | 8/Pk. |
£57.70
Pack of 8 |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||||
Description
Sample preparation and precise liquid handling in one device
A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.
- Concentrate and purify 1–100μL biological samples
- Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques
Order Info
Available in two packaging configurations: resealable trays (packs of 8 and 24 tips) and 96-place tip racks (packs of 96 and packs of 960 tips)
Specifications
10 μL | |
Universal Fit | |
ZipTip | |
Polypropylene |