missing translation for 'onlineSavingsMsg'
Learn More

Zirconium aluminide, 99% (metals basis), Thermo Scientific™

Product Code. 11308117
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11308117

Brand: Thermo Scientific Alfa Aesar 022886.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12004-83-0
Molecular Formula Al3Zr
Molecular Weight (g/mol) 172.169
MDL Number MFCD00064837
InChI Key PGLXYLYDLCVKEE-UHFFFAOYSA-N
Synonym zirconium aluminum, aluminum, compd. with zirconium 3:1, zirconium aluminide, trialumane zirconium
PubChem CID 6336544
IUPAC Name aluminum;zirconium
SMILES [Al].[Al].[Al].[Zr]

Specifications

Physical Form Lump
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 172.17
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Zirconium aluminide
compliance-icons
Product Identifier
  • Zirconium aluminide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.