missing translation for 'onlineSavingsMsg'
Learn More

Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals

Product Code. 15497365
250 g
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Zirconium Boride is used as an aerospace refractory, in cutting tools, and to protect thermocouple tubes. It is also acts as an antioxidant in carbon-bonded refractory ceramics, neutron absorber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12045-64-6
Molecular Formula B2Zr
Molecular Weight (g/mol) 112.844
MDL Number MFCD00064648
InChI Key VWZIXVXBCBBRGP-UHFFFAOYSA-N
PubChem CID 9812765
IUPAC Name boron;zirconium
SMILES [B].[B].[Zr]

Specifications

Melting Point 3000°C
Physical Form Powder
Quantity 250 g
Linear Formula ZrB2
UN Number UN3178
Solubility Information It is insoluble in water.
Formula Weight 112.84
Percent Purity 99.5%
Assay (metals basis excluding Hf)
Chemical Name or Material Zirconium boride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.