Learn More
Zirconium boride, 99.5% (metals basis excluding Hf), Thermo Scientific Chemicals
£90.05 - £310.55
Chemical Identifiers
CAS | 12045-64-6 |
---|---|
Molecular Formula | B2Zr |
Molecular Weight (g/mol) | 112.844 |
MDL Number | MFCD00064648 |
InChI Key | VWZIXVXBCBBRGP-UHFFFAOYSA-N |
Synonym | Zirconium diboride |
PubChem CID | 9812765 |
IUPAC Name | boron;zirconium |
SMILES | [B].[B].[Zr] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Preis | Menge & Verfügbarkeit | |||||
11356508
|
Thermo Scientific Alfa Aesar
012140.18 |
50 g |
£90.05
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346508
|
Thermo Scientific Alfa Aesar
012140.30 |
250 g |
£310.55
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Zirconium boride is use to improve resistance in zirconia-based, carbon-bonded refractories in contact with ferrous melts. Used in nuclear applications, aerospace refractory, in cutting tools and to protect thermocouple tubes. It also used in use of its relatively high conductivity, especially for a ceramic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
12045-64-6 | |
112.844 | |
VWZIXVXBCBBRGP-UHFFFAOYSA-N | |
9812765 | |
[B].[B].[Zr] |
B2Zr | |
MFCD00064648 | |
Zirconium diboride | |
boron;zirconium |
Spezifikation
3000°C | |
Powder | |
MFCD00064648 | |
Zirconium diboride | |
VWZIXVXBCBBRGP-UHFFFAOYSA-N | |
boron;zirconium | |
9812765 | |
99.5% | |
Zirconium boride |
12045-64-6 | |
B2Zr | |
UN3178 | |
Insoluble in water. | |
[B].[B].[Zr] | |
112.844 | |
112.84 | |
Plastic bottle |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 234-963-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.