missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
Zirconium Carbonate is used in the manufacturing of paint drier auxiliaries, pigments, water proof.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Physical Form | Powder |
Quantity | 2 kg |
Solubility Information | Insoluble in water. Soluble in hot concentrated acids. |
Formula Weight | 290.45 (Anhydrous) |
Density | 2.2 g/mL |
Chemical Name or Material | Zirconium carbonate, Basic hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'