missing translation for 'onlineSavingsMsg'
Learn More

Zirconium carbonate, basic hydrate, Thermo Scientific Chemicals

£53.87 - £154.77

Chemical Identifiers

CAS 57219-64-4
Molecular Formula CH2O7Zr2
Molecular Weight (g/mol) 308.47
MDL Number MFCD00172265
InChI Key ACWOHNQUPJAZIT-UHFFFAOYSA-J
Synonym hydrogen carbonate; zirconium 4+, zirconium 4+ ion tetrabicarbonate, zirconium iv carbonatebasic
PubChem CID 22645944
IUPAC Name [({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol
SMILES O[Zr](=O)OC(=O)O[Zr](O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11349338
View Documents
Thermo Scientific Alfa Aesar
043245.36
500 g
£53.87
500g
Please to purchase this item. Need a web account? Register with us today!
11359338
View Documents
Thermo Scientific Alfa Aesar
043245.A3
2 kg
£154.77
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zirconium Carbonate is used in the manufacturing of paint drier auxiliaries, pigments, water proof.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

57219-64-4
308.47
ACWOHNQUPJAZIT-UHFFFAOYSA-J
22645944
O[Zr](=O)OC(=O)O[Zr](O)=O
CH2O7Zr2
MFCD00172265
hydrogen carbonate; zirconium 4+, zirconium 4+ ion tetrabicarbonate, zirconium iv carbonatebasic
[({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol

Specifications

57219-64-4
Powder
MFCD00172265
Insoluble in water. Soluble in hot concentrated acids.
O[Zr](=O)OC(=O)O[Zr](O)=O
308.47
290.45 (Anhydrous)
Zirconium carbonate, Basic hydrate
White
CH2O7Zr2
hydrogen carbonate; zirconium 4+, zirconium 4+ ion tetrabicarbonate, zirconium iv carbonatebasic
ACWOHNQUPJAZIT-UHFFFAOYSA-J
[({[hydroxy(oxo)zirconio]oxy}carbonyl)oxy]zirconiumoylol
22645944
2.2 g/mL
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 260-633-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only