missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Zirconium Crucible

Brand:  Thermo Scientific™ 035986.KT

 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15493605

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

With SlideOn Ring; Ring/ Rim OD: 54mm
47mm
Zirconium Crucible
Zirconium
35mm
36mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers