Learn More
Zirconium dichloride oxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds an application as rubber additive and fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 400°C (decomposition) |
Physical Form | Lump |
Quantity | 250 g |
Assay Percent Range | 99.9%, metal basis |
Solubility Information | Soluble in water and alcohol. |
IUPAC Name | dichloro(oxo)zirconium |
Formula Weight | 178.13 (Anhydrous) |
Sensitivity | Hygroscopic |
Density | 1.91 |
Chemical Name or Material | Zirconium Dichloride Oxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X21K009 | Certificate of Analysis | 27/01/2025 | 086108.A1,086108.30,086108.18 |
S23G053 | Certificate of Analysis | 22/11/2022 | 086108.A1,086108.30,086108.18 |
Y10H075 | Certificate of Analysis | 12/04/2022 | 086108.A1,086108.30,086108.18 |
C16F48 | Certificate of Analysis | 28/12/2021 | 086108.30,078113.03,086108.18 |
K19T064 | Certificate of Analysis | 08/12/2021 | 086108.A1,086108.30,086108.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.