missing translation for 'onlineSavingsMsg'
Learn More

Zirconium Dichloride Oxide Hydrate, 99.9985% (Metal Basis), Thermo Scientific™, Puratronic™

Product Code. 11396148
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15461-27-5
Molecular Formula Cl2OZr
Molecular Weight (g/mol) 178.12
MDL Number MFCD00149898
InChI Key IPCAPQRVQMIMAN-UHFFFAOYSA-L
Synonym zirconyl chloride hydrate, zirconium dichloride oxide hydrate, zirconium, dichlorooxo-, monohydrate, zirconium oxychloride hydrate, cl2ozr.h2o, zirconium dichloride oxide hydrate, puratronic, zirconyl chloride hydrate trace metals basis
PubChem CID 16211499
SMILES Cl[Zr](Cl)=O

Specifications

Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.9985%, metal basis
Solubility Information Soluble in cold water,methanol,diethyl ether and alcohol.Slightly soluble in hydrochloric acid. Insoluble in organic solvents.
IUPAC Name oxozirconium;hydrate;dihydrochloride
Formula Weight 178.13 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Zirconium Dichloride Oxide Hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'