missing translation for 'onlineSavingsMsg'
Learn More
Zirconium fluoride, 99.5%, Thermo Scientific Chemicals Product Code.: 10792761

Zirconium fluoride, 99.5%, Thermo Scientific Chemicals

Product Code. 10792761
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10792761

Brand: Thermo Scientific Acros 299790250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-64-4
Molecular Formula F4Zr
Molecular Weight (g/mol) 167.21
MDL Number MFCD00011307

Specifications

Boiling Point 905.0°C
Melting Point 640.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 25 g
Assay Percent Range 99.4% min
Linear Formula ZrF4
Merck Index 15, 10375
Solubility Information Solubility in water: 1.388 g/100mL. Other solubilities: slightly soluble in hf
Formula Weight 167.21
Percent Purity 99.5%
Packaging Plastic bottle
Chemical Name or Material Zirconium fluoride, 99.5%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451552 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number299790250,299791000
Lot NumberA0391130 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number299790250,299791000
Lot NumberA0436634 Certificate TypeCertificate of Analysis Date28/11/2022 Catalog Number299790250,299791000
Lot NumberA0395061 Certificate TypeCertificate of Analysis Date22/03/2022 Catalog Number299790250,299791000
compliance-icons
Product Identifier
  • Zirconium fluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Zirconium fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium fluoride, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.