missing translation for 'onlineSavingsMsg'
Learn More
Zirconium foil, 0.025mm (0.001in) thick, annealed, 99.8% (metals basis excluding Hf), Thermo Scientific Chemicals Product Code.: 11368817

Zirconium foil, 0.025mm (0.001in) thick, annealed, 99.8% (metals basis excluding Hf), Thermo Scientific Chemicals

Product Code. 11368817
10 x 50 cm
Click to view available options
Quantity:
10 x 50 cm
10 x 200 cm
This item is not returnable. View return policy

Product Code. 11368817

Brand: Thermo Scientific Alfa Aesar 038369.VR

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Quantity 10 x 50 cm
Percent Purity 99.8%
Odor Odorless
Weight ≈8.3g/10x50cm
Form Foil
Assay (metals basis)
Chemical Name or Material Zirconium foil, 0.025mm (0.001 in.) thick, Annealed, Temper: soft

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY05G026 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number038369.VR,038369.VS
Lot NumberC24N59 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number038369.VR,038369.VS
Lot NumberX15F001 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number038369.VR,038369.VS
Lot NumberX15H057 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number038369.VR,038369.VS
Lot NumberF11W033 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number038369.VR,038369.VS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium foil, 0.025mm (0.001in) thick, annealed, 99.8% (metals basis excluding Hf), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.