missing translation for 'onlineSavingsMsg'
Learn More
Zirconium foil, 0.127mm (0.005in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 15445955

Zirconium foil, 0.127mm (0.005in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 15445955
125 x 2000 mm
Click to view available options
Quantity:
100 x 125 mm
125 x 500 mm
125 x 2000 mm
This item is not returnable. View return policy

Product Code. 15445955

Brand: Thermo Scientific Alfa Aesar 010594.IB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx.10.3g/100 x 125mm, Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Color Gray to Silver
Appearance Gray to silver foil
Assay Percent Range 99.5% (Metals basis)
Quantity 125 x 2000 mm
Odor Odorless
Form Foil
Chemical Name or Material Zirconium, Annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS16J064 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog Number010594,010594.IB,010594.GM,010594.GR
Lot NumberJ14G10 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010594.IB,010594.GM,010598.18,010594.GR
Lot NumberJ24G10 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010594.IB,010594.GM,010598.18,010594.GR
Lot NumberX26H039 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010594.IB,010594.GM,010598.18,010594.GR
Lot NumberM04I029 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010594.IB,010594.GM,010598.18,010594.GR

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium foil, 0.127mm (0.005in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.