missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) bromide, 99%, Thermo Scientific Chemicals

Product Code. 11334949
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11334949

Brand: Thermo Scientific Alfa Aesar 086139.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zirconium(IV) bromide is used as a starting material in the synthesis of zirconium hydride complexes as well as unique cluster compounds. It is also used as a biochemical for proteomics research and as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13777-25-8
Molecular Formula Br4Zr
Molecular Weight (g/mol) 410.84
MDL Number MFCD00049637
InChI Key LSWWNKUULMMMIL-UHFFFAOYSA-J
Synonym zirconium tetrabromide, zrbr4, zirconium iv bromide, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4, zirconium bromide zrbr4 8ci 9ci, zirconium iv bromide, anhydrous trace metals basis
PubChem CID 83727
IUPAC Name tetrabromozirconium
SMILES [Br-].[Br-].[Br-].[Br-].[Zr+4]

Specifications

Melting Point 450° (15 atm)
Physical Form Powder
Quantity 50 g
Assay Percent Range 99%
Solubility Information Soluble in water.
Formula Weight 410.86
Sensitivity Moisture sensitive
Density 4.201
Chemical Name or Material Zirconium(IV) Bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.