missing translation for 'onlineSavingsMsg'
Learn More
Zirconium(IV) chloride, Reactor Grade, 99.5+% (metals basis), Thermo Scientific Chemicals Product Code.: 11396458

Zirconium(IV) chloride, Reactor Grade, 99.5+% (metals basis), Thermo Scientific Chemicals

Product Code. 11396458
2 kg
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11396458

Brand: Thermo Scientific Alfa Aesar 012104.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Zirconium tetrachloride is commonly utilized as a Lewis acid catalyst in many organic synthesis reactions. It is an important source for organometallic complexes of zirconium. It is practiced in many areas like nuclear industry, metal industry, photography, glass, and ceramic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-11-6
Molecular Formula ZrCl4
MDL Number MFCD00011306
Synonym Zirconium tetrachloride

Specifications

Melting Point 331° (sublimation)
Physical Form Powder
Quantity 2 kg
Assay Percent Range >99.5% (metals basis)
Solubility Information Soluble in alcohol,ether,hydrochloric acid.
Formula Weight 233.03
Grade Reactor
Sensitivity Moisture sensitive
Density 2.803
Chemical Name or Material Zirconium(IV) chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS24K024 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number012104.09,012104.22,012104.A3,012104.36
Lot NumberQ15K044 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number012104.09,012104.22,012104.A3,012104.36
Lot NumberW30I027 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number012104.09,012104.22,012104.A3,012104.36
Lot NumberW30I026 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number012104.09,012104.22,012104.A3,012104.36
Lot NumberW16I009 Certificate TypeCertificate of Analysis Date23/09/2022 Catalog Number012104.09,012104.22,012104.A3,012104.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium(IV) chloride, Reactor Grade, 99.5+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.