Learn More
Zirconium(IV) n-propoxide, 70% w/w in n-propanol, Thermo Scientific Chemicals
£21.10 - £288.00
Chemical Identifiers
CAS | 23519-77-9 |
---|---|
Molecular Formula | C12H28O4Zr |
Molecular Weight (g/mol) | 327.58 |
MDL Number | MFCD00015307 |
InChI Key | XPGAWFIWCWKDDL-UHFFFAOYSA-N |
Synonym | zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt |
PubChem CID | 90139 |
SMILES | CCCO[Zr](OCCC)(OCCC)OCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11362598
|
Thermo Scientific Alfa Aesar
022989.18 |
50 g |
£21.10
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11372598
|
Thermo Scientific Alfa Aesar
022989.36 |
500 g |
£76.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382598
|
Thermo Scientific Alfa Aesar
022989.A3 |
2 kg |
£288.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zirconium(IV) n-propoxide is used as a precursor in the production of zirconia. It is also used to study the thermophysical properties like thermal conductivity and thermal effusivity of hybrid films and thereby adjusting optic properties of the films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23519-77-9 | |
327.58 | |
XPGAWFIWCWKDDL-UHFFFAOYSA-N | |
90139 |
C12H28O4Zr | |
MFCD00015307 | |
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt | |
CCCO[Zr](OCCC)(OCCC)OCCC |
Specifications
23519-77-9 | |
C12H28O4Zr | |
UN1993 | |
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt | |
XPGAWFIWCWKDDL-UHFFFAOYSA-N | |
propan-1-olate;zirconium(4+) | |
90139 | |
70% w/w in n-propanol | |
Moisture sensitive | |
Zirconium(IV) n-propoxide |
Liquid | |
MFCD00015307 | |
28°C (83°F) | |
Hydrolyzes in waterImmiscible with water. | |
CCCO[Zr](OCCC)(OCCC)OCCC | |
327.58 | |
327.56 | |
1.451 | |
1.044 g/mL |
Safety and Handling
GHS H Statement
H314-H226-H336
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c
H226-H318-H335-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 245-711-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only