missing translation for 'onlineSavingsMsg'
Learn More
Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals Product Code.: 10521451

Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals

Product Code. 10521451
1 kg, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10521451

Brand: Thermo Scientific Acros 190520010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs fillerShow MoreShow Less
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Boiling Point 4300.0°C
Melting Point 2700.0°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 98% min. (ZrO2+ HfO2)
Linear Formula ZrO2
Merck Index 15, 10380
Solubility Information Solubility in water: insoluble <1mg/l (20°C). Other solubilities: slightly soluble in hcl and hno3, slowly soluble in hf, disolves on heating with a mixt of 2 parts, h2so4 and 1 part h2o
Formula Weight 123.22
Percent Purity 98.5%
Packaging Glass bottle
Specific Gravity 5.6
Density 5.6000g/mL
Chemical Name or Material Zirconium(IV) oxide, 98.5%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0454552 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number190522500,190520010
Lot NumberA0453337 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number190522500,190520010
Lot NumberA0452035 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number190522500,190520010
Lot NumberA0450092 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number190522500,190520010
Lot NumberA0449026 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number190522500,190520010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium(IV) oxide, 98.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.