missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) oxide, 99.5% (metals basis excluding Hf), Hf <100ppm, Thermo Scientific™

£53.14 - £227.00

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359287
View Documents
Thermo Scientific Alfa Aesar
040140.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11349287
View Documents
Thermo Scientific Alfa Aesar
040140.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1314-23-4
123.22
MCMNRKCIXSYSNV-UHFFFAOYSA-N
62395
O=[Zr]=O
O2Zr
MFCD00011310 MFCD01868884
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
dioxozirconium

Specifications

∼5000°C
1314-23-4
Powder
MFCD00011310 MFCD01868884
zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
MCMNRKCIXSYSNV-UHFFFAOYSA-N
dioxozirconium
62395
99.5%
5.89 g/mL
Zirconium(IV) oxide
∼2700°C
White
O2Zr
14,10180
Soluble in alcohols,hydrofluoric and hot sulphuric acid. Insoluble in water.
O=[Zr]=O
123.22
123.22
Odorless
(metals basis excluding Hf), Hf <100ppm
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-227-2

missing translation for 'rtecsNumber' : ZH8800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications