Learn More
Zirconium(IV) oxide, calcia stabilized, 99.4% (metals basis excluding Hf), Thermo Scientific Chemicals
Description
The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | ≈5000°C |
Melting Point | ∽2700°C |
Physical Form | Powder |
Quantity | 250 g |
Solubility Information | Insoluble in water. soluble in HF,and hot Sulphuric acid. |
Formula Weight | 123.22 |
Percent Purity | 99.4% |
Packaging | Plastic bottle |
Chemical Name or Material | Zirconium(IV) oxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N11I025 | Certificate of Analysis | 12/04/2022 | 040452.A1,040453.18,040452.18,040452.30 |
N22I016 | Certificate of Analysis | 12/04/2022 | 040452.A1,040453.18,040452.18,040452.30 |
Y12G044 | Certificate of Analysis | 08/12/2021 | 040452.A1,040453.18,040452.18,040452.30 |
T15B047 | Certificate of Analysis | 07/12/2021 | 040452.A1,040453.18,040452.18,040452.30 |
J11M53 | Certificate of Analysis | 07/12/2021 | 040452.A1,040453.18,040452.18,040452.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.