missing translation for 'onlineSavingsMsg'
Learn More
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10356694

Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10356694
100 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10356694

Brand: Thermo Scientific Acros 427931000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23519-77-9, 71-23-8
Molecular Formula C12H28O4Zr
Molecular Weight (g/mol) 327.58
MDL Number MFCD00015307
InChI Key XPGAWFIWCWKDDL-UHFFFAOYSA-N
Synonym zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
PubChem CID 90139
SMILES CCCO[Zr](OCCC)(OCCC)OCCC

Specifications

Color Amber to Colorless
Density 1.0440g/mL
Flash Point 23°C
Assay Percent Range 27.0 to 29% (ZrO2)
Packaging AcroSeal™ Glass Bottle
Linear Formula Zr(OCH2CH2CH3)4
Quantity 100 mL
Specific Gravity 1.044
IUPAC Name zirconium(4+) tetrakis(propan-1-olate)
Formula Weight 327.57
Percent Purity ca. 70%
Physical Form Slightly Viscous Liquid
Chemical Name or Material Zirconium(IV) propoxide, ca. 70%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zirconium(IV) propoxide, solution in propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.