missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) sulfate tetrahydrate, 98+% (metals basis), Thermo Scientific Chemicals

Product Code. 11359457
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Catalyst support

Catalyst support.Zirconium disulfate tetrahydrate is used in organic synthesis. It is also used in tanning white leather. Further, it is used as catalyst supports to precipitate proteins and amino acids. It is also useful in chemical analysis. It plays an essential role as an additive for lubricants and as a pigment stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-31-3
Molecular Formula H8O12S2Zr
Molecular Weight (g/mol) 355.40
MDL Number MFCD00149267
InChI Key NLOQZPHAWQDLQW-UHFFFAOYSA-J
Synonym zirconium sulfate tetrahydrate, unii-os8l03qu95, sulfuric acid, zirconium 4+ salt 2:1 , tetrahydrate, zirconium iv sulfate tetrahydrate, zirconiumsulfatetetrahydrate, 2so4.zr.4h2o, ksc498a6d, zirconium 4+ tetrahydrate disulfate, zirconium 4+ ion tetrahydrate disulfate
PubChem CID 23219663
IUPAC Name zirconium(4+);disulfate;tetrahydrate
SMILES O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color White
Quantity 2 kg
Merck Index 14,10182
Solubility Information Soluble in water.
Formula Weight 355.40 (283.34 Anhydrous)
Percent Purity ≥98%
Density 3.22 g/mL
Assay (metals basis)
Chemical Name or Material Zirconium(IV) sulfate tetrahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'