missing translation for 'onlineSavingsMsg'
Learn More
Zirconium(IV) sulfate tetrahydrate, 98+% (metals basis), Thermo Scientific Chemicals Product Code.: 11369457

Zirconium(IV) sulfate tetrahydrate, 98+% (metals basis), Thermo Scientific Chemicals

Product Code. 11369457
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11369457

Brand: Thermo Scientific Alfa Aesar 041041.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Catalyst support

Catalyst support.Zirconium disulfate tetrahydrate is used in organic synthesis. It is also used in tanning white leather. Further, it is used as catalyst supports to precipitate proteins and amino acids. It is also useful in chemical analysis. It plays an essential role as an additive for lubricants and as a pigment stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-31-3
Molecular Formula H8O12S2Zr
Molecular Weight (g/mol) 355.40
MDL Number MFCD00149267
InChI Key NLOQZPHAWQDLQW-UHFFFAOYSA-J
Synonym zirconium sulfate tetrahydrate, unii-os8l03qu95, sulfuric acid, zirconium 4+ salt 2:1 , tetrahydrate, zirconium iv sulfate tetrahydrate, zirconiumsulfatetetrahydrate, 2so4.zr.4h2o, ksc498a6d, zirconium 4+ tetrahydrate disulfate, zirconium 4+ ion tetrahydrate disulfateShow MoreShow Less
PubChem CID 23219663
IUPAC Name zirconium(4+);disulfate;tetrahydrate
SMILES O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color White
Quantity 100 g
Merck Index 14,10182
Solubility Information Soluble in water.
Formula Weight 355.40 (283.34 Anhydrous)
Percent Purity ≥98%
Density 3.22 g/mL
Assay (metals basis)
Chemical Name or Material Zirconium(IV) sulfate tetrahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ02H004 Certificate TypeCertificate of Analysis Date27/12/2023 Catalog Number041041.22,041171.MD,041041.36,041041.A3
Lot NumberP30I028 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number041041.22,041171.MD,041041.36,041041.A3
Lot NumberM12I004 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number041041.22,041171.MD,041041.36,041041.A3
Lot NumberB24N19 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041041.22,041171.MD,041041.36,041041.A3
Lot NumberE10W030 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041041.22,041171.MD,041041.36,041041.A3
compliance-icons
Product Identifier
  • Zirconium(IV) sulfate tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Zirconium sulfate tetrahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium(IV) sulfate tetrahydrate, 98+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.