missing translation for 'onlineSavingsMsg'
Learn More
Zirconium(IV) sulfate tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11364898

Zirconium(IV) sulfate tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11364898
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11364898

Brand: Thermo Scientific Alfa Aesar 043194.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in catalysts, precipitation of amino acids and proteins, and tanning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-31-3
Molecular Formula H8O12S2Zr
Molecular Weight (g/mol) 355.40
MDL Number MFCD00149267
InChI Key NLOQZPHAWQDLQW-UHFFFAOYSA-J
Synonym zirconium sulfate tetrahydrate, unii-os8l03qu95, sulfuric acid, zirconium 4+ salt 2:1 , tetrahydrate, zirconium iv sulfate tetrahydrate, zirconiumsulfatetetrahydrate, 2so4.zr.4h2o, ksc498a6d, zirconium 4+ tetrahydrate disulfate, zirconium 4+ ion tetrahydrate disulfate
PubChem CID 23219663
IUPAC Name zirconium(4+);disulfate;tetrahydrate
SMILES O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Merck Index 14,10182
Solubility Information Soluble in water (525g/L).
Formula Weight 355.40 (283.34 Anhydrous)
Percent Purity 99.99%
Density 3.22 g/mL
Assay (metals basis)
Chemical Name or Material Zirconium (IV) sulfate tetrahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ14P13 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043194.22,043194.14
Lot NumberL12Y015 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043194.22,043194.14
Lot NumberD14W018 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043194.22,043194.14
Lot NumberY20G036 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043194.22,043194.14
Lot NumberW16H037 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043194.22,043194.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium(IV) sulfate tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.