missing translation for 'onlineSavingsMsg'
Learn More

Zirconium Oxide, Catalyst Support, Tungstated, Thermo Scientific™

£68.70 - £767.00

Chemical Identifiers

MDL Number MFCD00168096
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15452397
View Documents
Thermo Scientific Alfa Aesar
045597.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15462397
View Documents
Thermo Scientific Alfa Aesar
045597.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15472397
View Documents
Thermo Scientific Alfa Aesar
045597.A4
2.5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

MFCD00168096

Specifications

Pellets
MFCD00168096
Zirconium Oxide
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Zirconium oxide, catalyst support, tungstated

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Zirconium oxide

SDS
Documents

Documents

Product Certifications