Learn More
Zirconium oxide, polymeric precursor, Thermo Scientific Chemicals
Beschreibung
Zirconium oxide is used in precision ball valve balls and seats, rollers and guides for metal tube forming, powder compacting dies, marine pump seals and shaft guides, thread and wire guides, high temperature induction furnace susceptors, oxygen sensors and fuel cell membranes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Color | Yellow through Brown |
| Physical Form | Viscous Liquid |
| Quantity | 500 g |
| UN Number | UN1993 |
| Solubility Information | Soluble in alcohols |
| Odor | Alcohol-like |
| Sensitivity | Moisture sensitive |
| Chemical Name or Material | Zirconium oxide, Polymeric precursor |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.