missing translation for 'onlineSavingsMsg'
Learn More

Zirconium oxide, polymeric precursor, Thermo Scientific Chemicals

Product Code. 11345587
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Preparation of zirconium oxide films, solid electrolyte

Zirconium oxide is used in precision ball valve balls and seats, rollers and guides for metal tube forming, powder compacting dies, marine pump seals and shaft guides, thread and wire guides, high temperature induction furnace susceptors, oxygen sensors and fuel cell membranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Color Yellow through Brown
Physical Form Viscous Liquid
Quantity 100 g
UN Number UN1993
Solubility Information Soluble in alcohols
Odor Alcohol-like
Sensitivity Moisture sensitive
Chemical Name or Material Zirconium oxide, Polymeric precursor

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'