missing translation for 'onlineSavingsMsg'
Learn More
Zirconium oxide, yttria stabilized, polymeric precursor, Thermo Scientific™ Product Code.: 11349718

Zirconium oxide, yttria stabilized, polymeric precursor, Thermo Scientific™

Product Code. 11349718
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11349718

Brand: Thermo Scientific Alfa Aesar 044211.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

As a solid electrolyte and thermal barrier coating

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-23-4
Molecular Formula O2Zr
Molecular Weight (g/mol) 123.22
MDL Number MFCD00011310 MFCD01868884
InChI Key MCMNRKCIXSYSNV-UHFFFAOYSA-N
Synonym zirconia, zirconium oxide, zirconium oxide zro2, rhuligel, zirconium iv oxide, zirconium white, zirconic anhydride, pigment white 12, zirox zt 35, pcs filler
PubChem CID 62395
IUPAC Name dioxozirconium
SMILES O=[Zr]=O

Specifications

Physical Form Liquid
Quantity 5 g
UN Number UN1993
Solubility Information Soluble in hexane, toluene, alcohol
Chemical Name or Material Zirconium oxide, Yttria stabilized, polymeric precursor

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ27F011 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number044210.36,044211.06,044210.22,044210.A3,044211.03
Lot NumberS11G012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044210.36,044211.06,044210.22,044210.A3,044211.03
Lot NumberJ05P17 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044210.36,044211.06,044210.22,044210.A3,044211.03
Lot Number61000376 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044210.36,044211.06,044210.22,044210.A3,044211.03
Lot NumberG01P16 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044210.36,044211.06,044210.22,044210.A3,044211.03
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium oxide, yttria stabilized, polymeric precursor, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.