missing translation for 'onlineSavingsMsg'
Learn More

Zirconium oxide Yttrium oxide substrate, 10x10x1mm, polished one side, 100 orientation, Thermo Scientific Chemicals

£78.99 - £322.61

Specifications

Chemical Name or Material Zirconium Oxide Yttrium Oxide Substrate
Name Note Polished One Side
Physical Form Plate
Molecular Formula ZrO2 + 9.5% Y2O3
MDL Number MFCD01868885
View More Specs

Produits 2
Code produit Marque Quantity Prix Quantité et disponibilité  
Code produit Marque Quantity Prix Quantité et disponibilité  
11308867
missing translation for 'viewDocuments' Détails de la promotion
Thermo Scientific Alfa Aesar
038509.KT
1 Ea.
£78.99
Each
Please to purchase this item. Need a web account? Register with us today!
11398857
missing translation for 'viewDocuments' Détails de la promotion
Thermo Scientific Alfa Aesar
038509.KU
5 Ea.
£322.61
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécification

Spécification

Zirconium Oxide Yttrium Oxide Substrate
Plate
MFCD01868885
Polished One Side
ZrO2 + 9.5% Y2O3
Odorless
Vidéos
Sécurité et traitement

Sécurité et traitement

missing translation for 'einecsNumber' : 215-227-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Certificats

RUO – Research Use Only