missing translation for 'onlineSavingsMsg'
Learn More
Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals Product Code.: 11388288

Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals

Product Code. 11388288
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11388288

Brand: Thermo Scientific Alfa Aesar 039490.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for glass, optic and ceramic applications, advanced electronics and in light weight structural components in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-70-3
MDL Number MFCD00210648
Synonym zirconium titanium oxide, tetrahydrate titanium zirconium

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 203.11
Percent Purity 99.5%
Odor Odorless
Assay (metals basis excluding Hf), Hf <75ppm
Chemical Name or Material Zirconium titanium oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL27F28 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039490.22,039493.04,039490.14
Lot NumberD14R016 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039490.22,039493.04,039490.14
Lot NumberI21Y019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039490.22,039493.04,039490.14
Lot NumberE27U040 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039490.22,039493.04,039490.14
Lot NumberJ08H24 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number039490.22,039493.04,039490.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium titanium oxide, 99.5% (metals basis excluding Hf), Hf <75ppm, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.