missing translation for 'onlineSavingsMsg'
Learn More
Zirconium wire, 0.127mm (0.005in) dia, 99.95% (metals basis excluding Hf), Hf nominal 0.25%, Thermo Scientific Chemicals Product Code.: 11336557

Zirconium wire, 0.127mm (0.005in) dia, 99.95% (metals basis excluding Hf), Hf nominal 0.25%, Thermo Scientific Chemicals

Product Code. 11336557
5 m
Click to view available options
Quantity:
1 m
5 m
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11336557

Brand: Thermo Scientific Alfa Aesar 000419.G5

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Form: Approx.0.082g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Color Gray to Silver
Appearance Gray to silver wire
Density 6.506g/cm3
Assay Percent Range 99.95% (Metals basis excluding Hf)
Quantity 5 m
Odor Odorless
Form Wire
Chemical Name or Material Zirconium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS16I067 Certificate TypeCertificate of Analysis Date22/08/2022 Catalog Number000419.G5,000419.G1
Lot NumberS16I066 Certificate TypeCertificate of Analysis Date22/08/2022 Catalog Number000419.G5,000419.G1
Lot NumberR27I009 Certificate TypeCertificate of Analysis Date18/08/2022 Catalog Number000419.G5,000419.G1
Lot NumberZ30H022 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number000419.G5,000419.G1
Lot NumberP22H053 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000419.G5,000419.G1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium wire, 0.127mm (0.005in) dia, 99.95% (metals basis excluding Hf), Hf nominal 0.25%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.