missing translation for 'onlineSavingsMsg'
Learn More
Zirconium wire, 0.5mm (0.020in) dia, annealed, 99.2% (metals basis excluding Hf) Hf 4.5% max, Thermo Scientific Chemicals Product Code.: 15454935

Zirconium wire, 0.5mm (0.020in) dia, annealed, 99.2% (metals basis excluding Hf) Hf 4.5% max, Thermo Scientific Chemicals

Product Code. 15454935
1 m
Click to view available options
Quantity:
1 m
5 m
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15454935

Brand: Thermo Scientific Alfa Aesar 045056.G1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Quantity 1 m
Percent Purity 99.2%
Odor Odorless
Weight ≈1.31g/m
Form Wire
Assay (metals basis excluding Hf) Hf 4.5% max
Chemical Name or Material Zirconium wire, 0.5mm (0.020 in.) dia., Annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK15W040 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045056.G1,045056.G5,045057.G9
Lot NumberH21Y019 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045056.G1,045056.G5,045057.G9
Lot NumberU18A013 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045056.G1,045056.G5,045057.G9
Lot NumberZ01B013 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045056.G1,045056.G5,045057.G9
Lot NumberS26E043 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045056.G1,045056.G5,045057.G9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zirconium wire, 0.5mm (0.020in) dia, annealed, 99.2% (metals basis excluding Hf) Hf 4.5% max, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.